Orders LinkContactUsLinkComponent
Sample Containers, Polystyrene, Greiner Bio-One
Supplier:   GREINER BIO-ONE
Sample Containers, Polystyrene, Greiner Bio-One
Supplier:  GREINER BIO-ONE
Certificates
Sample Containers, Polystyrene, Greiner Bio-One
Sample Containers, Polystyrene, Greiner Bio-One
2 Options Available Below
Product Details & Documents
This collection of clear, polystyrene sample containers is designed to meet general-purpose collection, storage, and transport requirements for most lab environments. Containers are available with flat- or conical-bottom styles, and can be supplied sterile or nonsterile. Flat-bottom containers are supplied with plastic or metal screw caps, and conical-bottom containers are supplied with plastic, wadded screw caps.
  • Clear polystyrene construction
  • Supplied sterile or nonsterile
  • Flat or conical bottom styles
  • Supplied with scrap caps
Containers are available with flat- or conical-bottom styles, and can be supplied sterile or nonsterile. Flat-bottom containers are supplied with plastic or metal screw caps, and conical-bottom containers are supplied with plastic, wadded screw caps.
Product Family Options
Product Information
  • Capacity
    Height
    Width
    Label Type
    Sterility
    Availability
    Price
  • 89085-340
    Flat-Bottom Container, with Metal Screw Cap
    Flat-Bottom Container, with Metal Screw Cap
    100 ml (3.4 oz.)
    78 mm
    48 mm
    No label
    Non sterile
    Log in for availability
    Log in for availability
    Case (160UN)
    Case (160UN) - $7,240.00
    $7,240.00
    Case (160UN)
       $7,240.00
    Supplier Number: 224170
  • 89085-342
    Flat-Bottom Container, with Metal Screw Cap
    Flat-Bottom Container, with Metal Screw Cap
    148 ml (5 oz.)
    107 mm
    49 mm
    Plain
    Non sterile
    Log in for availability
    Log in for availability
    Case (120UN)
    Case (120UN) - $7,030.00
    $7,030.00
    Case (120UN)
       $7,030.00
    Supplier Number: 225180