Especificaciones
- Filter Media:Glass Fiber/Bio-Inert® Nylon
- Tamaño de poro:3.0 µm/ 0.2 µm
- Well Volume:1 ml
- Referencia:28148-626
- Working Volume:900 µl
- Nº de proveedor:5053
Especificaciones
Acerca de este artículo
Constructed of chemically resistant, biologically inert polypropylene, plates are optimized for a variety of applications requiring larger per well volume processing.
- Chemically resistant material
- Extended flow directors
- Individually sealed wells
Plates feature extended flow directors to eliminate splattering and crosstalk of filtrate, a serialized bar code label for use in automated tracking systems, robotics-friendly design, standard SBS format, and individually sealed wells to eliminate crosstalk.
Available in a variety of membranes and membrane configurations to meet various application needs. Choose from GHP (hydrophilic polypropylene), glass fiber (borosilicate glass without binder), glass fiber over Bio-Inert® modified low protein-binding nylon, hydrophobic PTFE, Supor® polyethersulfone, or Omega™ polyethersulfone (PES) membranes.