Orders LinkContactUsLinkComponent
VWR® Polypropylene Specimen Containers
VWR® Polypropylene Specimen Containers
VWR® Polypropylene Specimen Containers
Supplier:  VWR International
Certificates
VWR® Polypropylene Specimen Containers
VWR® Polypropylene Specimen Containers
3 Options Available Below
Product Details & Documents
Sturdy, shatter-resistant containers are manufactured from transparent polypropylene to allow constant sample viewing. Containers feature molded graduations in 10mL and 0.5oz. increments. Polyethylene snap-on lids are sold separately, except where noted. Lids create a secure seal, and offer space for date, name, doctor, and room number molded into the surface. Components are available in nonsterile or sterile formats.
  • Sturdy, shatter-resistant design
  • Transparent siding
  • Polypropylene construction
  • Available in nonsterile or sterile formats
Containers feature molded graduations in 10 mL and 0.5 oz. increments. Polyethylene snap-on lids are sold separately, except where noted. Lids create a secure seal, and offer space for date, name, doctor, and room number molded into the surface. Components are available in nonsterile or sterile formats.
Ordering information: Lids fit both 5 and 8 oz. containers. Bulk items are packaged 20 per bag, 25 bags per case.
Product Family Options
Product Information
  • Capacity
    Dimensions
    Packaging
    Sterility
    Availability
    Price
  • Container
    148 ml (5 oz.)
    73.7 Dia x 66.3 H mm
    Bulk
    Sterile
    Log in for availability
    Log in for availability
    Case (500UN)
    Case (500UN) - $4,695.77
    $4,695.77
    Case (500UN)
       $4,695.77
  • 89125-844
    Container with Lid
    Container with Lid
    148 ml (5 oz.)
    73.7 Dia x 66.3 H mm
    Individually Wrapped
    Sterile
    Discontinued
  • Container
    237 ml (8 oz.)
    73.7 Dia x 100.5 H mm
    Bulk
    Sterile
    Log in for availability
    Log in for availability
    Case (500UN)
    Case (500UN) - $5,172.44
    $5,172.44
    Case (500UN)
       $5,172.44
  • 89125-846
    Container with Lid
    Container with Lid
    237 ml (8 oz.)
    73.7 Dia x 100.5 H mm
    Individually Wrapped
    Sterile
    Log in for availability
    Log in for availability
    Case (100UN)
    Case (100UN) - $3,020.90
    $3,020.90
    Case (100UN)
       $3,020.90