ContactUsLinkComponent
Flip-Top Containers with Lock Seal, Capitol Vial®
Flip-Top Containers with Lock Seal, Capitol Vial®
Catálogo # 46610-586
undefined
Flip-Top Containers with Lock Seal, Capitol Vial®
Catálogo # 46610-586
Número de proveedor:  04HPLS
Especificaciones
  • Ø int.:
    43 mm (111/16")
  • Capacidad:
    120 ml (4 oz.)
  • Descripción:
    High Profile
  • Altura:
    90 mm (3½")
  • Referencia:
    46610-586
  • Nº de proveedor:
    04HPLS
Especificaciones
Acerca de este artículo
Triple-seal containers are sterile, airtight, and leakproof. Nonabsorptive, nonaerosoling containers are made of FDA-approved, food grade 100% polypropylene. Hinged top enables opening and closing with one hand. Ideal for storing and packaging a wide range of solids and liquids in applications such as medical device packaging, reagent packaging, solid sample storage, and water testing. Containers feature a lock seal tab that allows the user to lock the lid to the base of the container.
Unique process physically closes containers in the mold during manufacture to ensure an airtight seal and zero particulates >0.5µ. Process also produces sterile containers that do not require a second means of sterilization for most applications. Color: clear.
Nonabsorptive, nonaerosoling containers are made of FDA-approved, food grade 100% polypropylene. Hinged top enables opening and closing with one hand. Ideal for storing and packaging a wide range of solids and liquids in applications such as medical device packaging, reagent packaging, solid sample storage, and water testing. Containers feature a lock seal tab that allows the user to lock the lid to the base of the container.
Certificaciones: Containers conform to FDA and SAMHSA specifications.