CHEMSTAR™ Vacuum Pumps, Welch®
Catalog # 54994-050
Supplier: WELCH VACUUM BY GARDNER DENVER
Product available on GSA Advantage®
Specifications
- Descripción:Pump, ISO flange NW16
- Dimensiones:45.2 x 22.9 x 32 cm (17.8 x 9 x 12.6")
- Referencia:54994-050
- Detalles eléctricos:115 V, 60 Hz
- Peso al enviarse:30.8 kg (68 lbs.)
- Se ajusta al D.I. de la tubería:11.1 mm (⁷⁄₁₆")
- HP:1/3
- Capacidad de aceite:0.59 L (0.62 qts.)
- Nº de proveedor:1400N-01
- rpm:580
Specifications
About this item
Corrosive gases, such as HCl, HBr, Br2, Cl2, and F3CCO2H, are commonly used for peptide chemistry, molecular biology, and biochemistry applications
CHEMSTAR™ pumps resist corrosion and handle corrosive gases to pressures below 1 torr. They feature nitrogen purge capability, high contamination tolerance, low speeds for low wear, and inert components. CHEMSTAR™ pumps have an ultimate pressure of 1 x 10-4 torr. Contoured, molded belt-guard and base on 54994-052 and 54993-550 are uniquely designed for quieter operation and prevent visible oil weeping at seal.