DUOSEAL® Belt-Drive Vacuum Pumps, Single Stage, Welch®
Catalog # 54972-709
Supplier: WELCH VACUUM BY GARDNER DENVER
Product available on GSA Advantage®
Specifications
- Dimensiones:45 x 22.8 x 25.7 mm (1723/32 x 9 x 10")
- Referencia:54972-709
- Detalles eléctricos:115 V, 60 Hz, 1 phase
- Peso al enviarse:23.1 kg (50 lbs.)
- Libre desplazamiento de aire:35 L/min (1.2 cfm)
- HP:1/3
- Capacidad de aceite:0.47 L (0.5 qts.)
- Tubing I.D.:11.1 mm (⁷⁄₁₆")
- Nº de proveedor:1399B-01
- rpm:750
Specifications
About this item
Pumps are ideal for continuous-duty applications, including vacuum distillations, glove box transfer chamber, residue solvent removal, and resin degassing
They feature thermal overload protection standard and operate at a low rpm which allows them to reach a low ultimate pressure (0.02mm Hg). Operating at low speed also reduces the rate of friction and wear on pump components which ensures a long service life.
Ordering information: Exhaust filters are available separately and are supplied with housing and element.