Orders LinkContactUsLinkComponent
Falcon® Sample Containers, Polypropylene, Sterile, Corning®
Falcon® Sample Containers, Polypropylene, Sterile, Corning®
220 ml (7.4 oz.), Containers with Lid, Individually Wrapped
Catalog # 15704-084
Supplier:   Corning
Número de proveedor #:  354015
Falcon® Sample Containers, Polypropylene, Sterile, Corning®
Catalog # 15704-084
Supplier:  Corning
Falcon® Sample Containers, Polypropylene, Sterile, Corning®
Falcon® Sample Containers, Polypropylene, Sterile, Corning®
220 ml (7.4 oz.), Containers with Lid, Individually Wrapped
Price
$4,570.00
Log in Para disponibilidad
Case (100UN)
Product Details & Documents
Sterile, disposable polypropylene containers provide secure sample containment. Inert and chemically resistant to commonly used laboratory reagents at room temperature. Containers feature molded graduations in milliliters and ounces.
  • Molded-in graduations for easy measurement
  • Inert and chemically resistant to commonly used laboratory reagents at room temperature
Convenient, single-use containers for collection, transport and storage of a wide variety of specimens.
Polyethylene lid is frosted for easy marking, and fits both container sizes.
Especificaciones
  • Capacity:
    220 ml (7.4 oz.)
  • Description:
    Containers with Lid
  • Packaging:
    Individually Wrapped