Orders LinkContactUsLinkComponent
Sanitary Counter Brushes, Perfex
PP, White, Plastic, 14", 2¼", 1⅝"
  22940-024
 :  3050W
Sanitary Counter Brushes, Perfex
  22940-024
Sanitary Counter Brushes, Perfex
Sanitary Counter Brushes, Perfex
PP, White, Plastic, 14", 2¼", 1⅝"
$556.27
Each (1UN)
Sanitary Counter Brushes quickly remove fine dust, dirt and other debris from workstations, counter tops, and shelves without scattering particles.
  • Ideal for a variety of clean-up chores
  • Durable fiber maintains shape and is visually coded to isolate areas of use
  • Poly block and fiber will not absorb bacteria, liquids or odors like nylon and natural fibers
  • Unaffected by water, grease, petroleum products, detergent, sanitizer and solvents
Medium-texture polypropylene fiber is heat-fused into impact-resistant polymer brush block, eliminating premature fiber fallout and areas for contaminants to collect.
  • Bristle Color:
    White
  • Bristle Material:
    PP
  • Handle Material:
    Plastic
  • Total length:
    14"
  • Width:
    1⅝"
  • Trim Length:
    2¼"