XPert™ Balance Enclosures, Labconco®
Acerca de este artículo
These low-profile enclosures provide user protection by keeping powders, particulates, and fumes contained during weighing procedures
Sturdy glacier white and gray, dry powder epoxy-coated aluminum frame and steel rear plenum and baffle dissipate static charge. Tempered safety glass front sash, sides, and top offer excellent visibility, ambient light, and protection. All models with interior heights of 56.5cm (221/4") have an operating sash height of 20.3cm (8"). The 0.6m (2'), 0.9m (3'), and 1.2m (4') models with interior heights of 80cm (311/2") have an operating sash height of 25.4cm (10"); the 1.5m (5'), 1.8m (6'), and 2.4m (8') models with interior heights of 80cm (311/2") have an operating sash height of 30.5cm (12"). Conservative operating sash heights limit exhaust demand, conserve tempered room air, and require a smaller remote blower than most fume hoods. Ergonomic angled sash allows a closer view, reduces glare, and provides a more comfortable operating position than vertical sashes. The hinged sash pivots upward and locks to a loading height of 48.3cm (19"). Models with 25.4cm (10") and 30.5cm (12") operating sash heights open to 70.4cm (2711/16") for loading. The 1.5m (5'), 1.8m (6'), and 2.4m (8') models incorporate a gas-assist lift. Two utility ports allow passage of tubing and cords. Enclosures feature 5.1x25.4cm (2x10") rectangular exhaust collars on the top and bottom for connection to ductwork or portable exhauster.
Enclosures also feature an optional built-in airflow monitor. If the hood’s face velocity falls below the setpoint for more than six seconds, an audible alarm sounds and a red light glows. The alarm can be muted. On the Guardian™ monitor, an LED glows green when the airflow is within setpoint range. The Guardian 1000 digital monitor features an LCD display of actual face velocity and an RS-232 port. Remote blowers 30153-980, 30153-982, and 82010-730 have a 1/4hp, totally enclosed, fan-cooled (TEFC) standard motor capable of overcoming external static pressure loss of 0.5in.H2O at 420cfm. Housing and wheel are made of corrosion-resistant, phenolic-coated steel. Blower inlet has an I.D. of 15.2cm (6"). Outlet dimensions are 10.8Wx18.7Lcm (41/4x73/8"). Applications involving high concentrations of solvents may require the explosion-proof blower. Stainless steel work surfaces feature a spill trough. The utility shelf kit has three epoxy-coated steel shelves that can hold a printer, utensils, and other miscellaneous supplies. Shelves attach to the rear baffle perforations of the enclosure.
The 59.4cm (2313/32") deep interior easily accommodates large micro and analytical balances. The unique Clean-Sweep™ airfoil allows air to sweep the work surface for maximum containment. Airflow openings pull air under the airfoil, creating a constant barrier of protection from contaminants. The upper containment sash foil bleeds air into the enclosure to direct concentrations of contaminants away from the operator’s breathing zone. The upper dilution air supply introduces room air at the top of the sash to dilute contaminants in the upper chamber and bathes the back of the sash with clean air. The perforated rear baffle promotes horizontal laminar airflow to maximize containment. Side-entry airfoils allow air to sweep across the interior glass surfaces to enhance containment.
Certifications: All enclosures conform to modified ASHRAE 110-1995 standards, SEFA 1-2002 standards, and ANSI Z9.5-1-993 standards. The 115V models are ETL and ETL-C listed. The 230V models are CE marked.
Ordering information: Items designated below as REDISHIP can be shipped within 48 hours of receiving your order. Enclosures must be connected to either one or more FilterMate™ portable exhauster (see 82010-632 series), house exhaust, or a remote blower. An optional work surface is also recommended. The 1.5m (5'), 1.8m (6'), and 2.4m (8') models with 90.2cm (351/2") depth include a left side-mounted waste chute that accepts bags 25.4x40.6cm (10x16") and larger. For required base cabinets, see the Protector® acid storage cabinets (46610-412 series), solvent storage cabinets (46610-440 series), and standard cabinets (46610-404 series). To add the optional Guardian™ Jr. and Guardian™ 1000 airflow monitors 82010-702 and 82010-706 at a later time, see 82007-280 series. Blowers, exhaust dampers, transition adapters, and gray work surfaces are also available; contact your VWR sales representative for more information.
3 Options Available Below
Product Details & Documents
These low-profile enclosures provide user protection by keeping powders, particulates, and fumes contained during weighing procedures
Sturdy glacier white and gray, dry powder epoxy-coated aluminum frame and steel rear plenum and baffle dissipate static charge. Tempered safety glass front sash, sides, and top offer excellent visibility, ambient light, and protection. All models with interior heights of 56.5cm (221/4") have an operating sash height of 20.3cm (8"). The 0.6m (2'), 0.9m (3'), and 1.2m (4') models with interior heights of 80cm (311/2") have an operating sash height of 25.4cm (10"); the 1.5m (5'), 1.8m (6'), and 2.4m (8') models with interior heights of 80cm (311/2") have an operating sash height of 30.5cm (12"). Conservative operating sash heights limit exhaust demand, conserve tempered room air, and require a smaller remote blower than most fume hoods. Ergonomic angled sash allows a closer view, reduces glare, and provides a more comfortable operating position than vertical sashes. The hinged sash pivots upward and locks to a loading height of 48.3cm (19"). Models with 25.4cm (10") and 30.5cm (12") operating sash heights open to 70.4cm (2711/16") for loading. The 1.5m (5'), 1.8m (6'), and 2.4m (8') models incorporate a gas-assist lift. Two utility ports allow passage of tubing and cords. Enclosures feature 5.1x25.4cm (2x10") rectangular exhaust collars on the top and bottom for connection to ductwork or portable exhauster.
Enclosures also feature an optional built-in airflow monitor. If the hood’s face velocity falls below the setpoint for more than six seconds, an audible alarm sounds and a red light glows. The alarm can be muted. On the Guardian™ monitor, an LED glows green when the airflow is within setpoint range. The Guardian 1000 digital monitor features an LCD display of actual face velocity and an RS-232 port. Remote blowers 30153-980, 30153-982, and 82010-730 have a 1/4hp, totally enclosed, fan-cooled (TEFC) standard motor capable of overcoming external static pressure loss of 0.5in.H2O at 420cfm. Housing and wheel are made of corrosion-resistant, phenolic-coated steel. Blower inlet has an I.D. of 15.2cm (6"). Outlet dimensions are 10.8Wx18.7Lcm (41/4x73/8"). Applications involving high concentrations of solvents may require the explosion-proof blower. Stainless steel work surfaces feature a spill trough. The utility shelf kit has three epoxy-coated steel shelves that can hold a printer, utensils, and other miscellaneous supplies. Shelves attach to the rear baffle perforations of the enclosure.
The 59.4cm (2313/32") deep interior easily accommodates large micro and analytical balances. The unique Clean-Sweep™ airfoil allows air to sweep the work surface for maximum containment. Airflow openings pull air under the airfoil, creating a constant barrier of protection from contaminants. The upper containment sash foil bleeds air into the enclosure to direct concentrations of contaminants away from the operator’s breathing zone. The upper dilution air supply introduces room air at the top of the sash to dilute contaminants in the upper chamber and bathes the back of the sash with clean air. The perforated rear baffle promotes horizontal laminar airflow to maximize containment. Side-entry airfoils allow air to sweep across the interior glass surfaces to enhance containment.
Certifications: All enclosures conform to modified ASHRAE 110-1995 standards, SEFA 1-2002 standards, and ANSI Z9.5-1-993 standards. The 115V models are ETL and ETL-C listed. The 230V models are CE marked.
Ordering information: Items designated below as REDISHIP can be shipped within 48 hours of receiving your order. Enclosures must be connected to either one or more FilterMate™ portable exhauster (see 82010-632 series), house exhaust, or a remote blower. An optional work surface is also recommended. The 1.5m (5'), 1.8m (6'), and 2.4m (8') models with 90.2cm (351/2") depth include a left side-mounted waste chute that accepts bags 25.4x40.6cm (10x16") and larger. For required base cabinets, see the Protector® acid storage cabinets (46610-412 series), solvent storage cabinets (46610-440 series), and standard cabinets (46610-404 series). To add the optional Guardian™ Jr. and Guardian™ 1000 airflow monitors 82010-702 and 82010-706 at a later time, see 82007-280 series. Blowers, exhaust dampers, transition adapters, and gray work surfaces are also available; contact your VWR sales representative for more information.
Documents
Labconco REDISHIP Literature 2021
Product Literature
Specifications for Product Family
Especificaciones
- Catalog No:
- 89004-458
- 89004-462
- 89004-466
- Model:
- 2' Wide Enclosures
- 3' Wide Enclosures
- 4' Wide Enclosures
- Exhaust volume:
- 120 cfm (at 100 fpm), 95 cfm (at 80 fpm), 70 cfm (at 60 fpm)
- 185 cfm (at 100 fpm), 145 cfm (at 80 fpm), 110 cfm (at 60 fpm)
- 250 cfm (at 100 fpm), 200 cfm (at 80 fpm), 150 cfm (at 60 fpm)
- Static pressure loss:
- 0.05 in. H2O (at 100 fpm), 0.03 in. H2O (at 80 fpm), 0.02 in. H2O (at 60 fpm)
- 0.10" H2O (at 100 fpm), 0.06" H2O (at 80 fpm), 0.04" H2O (at 60 fpm)
- 0.16 in. H2O (at 100 fpm), 0.10 in. H2O (at 80 fpm), 0.06 in. H2O (at 60 fpm)
Especificaciones
Product Family Options
Información del producto
- WidthInside DepthInside HeightInside WidthOutside DepthOutside HeightOutside WidthREDISHIPAirflow MonitorShipping WeightAvailabilityPrice
- 2 ft.23⅜"221/4"22¾"29"22¹³⁄₁₆"24"YesNo48 kg (105 lbs.)Log in Para disponibilidadEach (1UN)$86,045.00Supplier Number: 3930200Especificaciones:Product available on GSA Advantage®
More
Specifications
Cat. No.89004-458Model2' Wide EnclosuresExhaust volume120 cfm (at 100 fpm), 95 cfm (at 80 fpm), 70 cfm (at 60 fpm)Static pressure loss0.05 in. H2O (at 100 fpm), 0.03 in. H2O (at 80 fpm), 0.02 in. H2O (at 60 fpm)Product available on GSA Advantage® - Supplier Number: 3930300Especificaciones:Product available on GSA Advantage®
More
Specifications
Cat. No.89004-462Model3' Wide EnclosuresExhaust volume185 cfm (at 100 fpm), 145 cfm (at 80 fpm), 110 cfm (at 60 fpm)Static pressure loss0.10" H2O (at 100 fpm), 0.06" H2O (at 80 fpm), 0.04" H2O (at 60 fpm)Product available on GSA Advantage® - 4 ft.23⅜"221/4"46¾"29"22¹³⁄₁₆"48"YesNo73 kg (160 lbs.)Log in Para disponibilidadEach (1UN)$213,142.88Supplier Number: 3930400Especificaciones:Product available on GSA Advantage®
More
Specifications
Cat. No.89004-466Model4' Wide EnclosuresExhaust volume250 cfm (at 100 fpm), 200 cfm (at 80 fpm), 150 cfm (at 60 fpm)Static pressure loss0.16 in. H2O (at 100 fpm), 0.10 in. H2O (at 80 fpm), 0.06 in. H2O (at 60 fpm)Product available on GSA Advantage®
Accessories
Información del producto
- Catalog #REDISHIPShipping WeightAvailabilityPrice
- Upper Exhaust Transition Adapter, 6" O.D. DuctUpper Exhaust Transition Adapter, 6" O.D. Duct82010-718Yes5 lbs.Log in Para disponibilidadEach (1UN) - $4,437.50
Información del producto
- Catalog #REDISHIPShipping WeightAvailabilityPrice
- Lower Exhaust Transition Adapter, 6" O.D. DuctLower Exhaust Transition Adapter, 6" O.D. Duct82010-722Yes5 lbs.Log in Para disponibilidadEach (1UN) - $4,437.50
Información del producto
- Catalog #Inside DepthOutside HeightAvailabilityPrice
- Solid Epoxy Dished Work Surface
Información del producto
- Catalog #REDISHIPShipping WeightAvailabilityPrice
- Solid Epoxy Dished Work Surface for 3' ModelsSolid Epoxy Dished Work Surface for 3' Models82010-692Yes72 lbs.Log in Para disponibilidadEach (1UN) - $24,677.87