ContactUsLinkComponent
Corning® 96-Well Polypropylene Storage Blocks
  29445-130
 :  Corning
Product available on GSA Advantage®
undefined
Corning® 96-Well Polypropylene Storage Blocks
  29445-130
 :  Corning
 :  3959
Product available on GSA Advantage®
  • Esterilidad:
    Non sterile
  • Well Volume:
    1 mL
  • Color:
    Clear
  • Descripción:
    Storage block, 96-well, natural, round well
  • Referencia:
    29445-130
  • Well Shape:
    Round-Bottom
  • Nº de proveedor:
    3959
Plates are available in V-bottom and round-bottom formats to meet a wide variety of assay and storage needs.
Robolid combines the sealing ability of a storage mat with the rigidity of a plastic lid and is designed for repeated application and removal by automation and to prevent short-term evaporation. Robolid also features silicone sealing plugs for organic solvent resistance and low extractables.
All sterile products are certified RNase- and DNase-free. Built to the industry standard footprint.
 : Storage mats and sealing tapes sold separately.