Orders LinkContactUsLinkComponent
96-Well Polypropylene Tubes, Corning®
96-Well Polypropylene Tubes, Corning®
Supplier:   Corning
96-Well Polypropylene Tubes, Corning®
Supplier:  Corning
Certificates
96-Well Polypropylene Tubes, Corning®
96-Well Polypropylene Tubes, Corning®
3 Options Available Below
Product Details & Documents
Cluster tube system is composed of ninety-six 1.2mL polypropylene tubes arranged in a microtitration plate format. Tubes are available individually or in strips of eight tubes.
  • Composed of 96 PP tubes in a standard microplate format
  • 1.2 ml tubes available individually or in strips of 8 tubes
  • PE tube caps available in 8-cap strips
Packaging: Supplied bagged or racked.
Product Family Options
Información del producto
  • Capacity
    Packaging
    Sterility
    Availability
    Price
  • Individual Tubes
    1.2 ml
    Bulk
    Non sterile
    Log in Para disponibilidad
    Log in Para disponibilidad
    Case (960UN)
    Case (960UN) - $2,330.00
    $2,330.00
    Case (960UN)
       $2,330.00
    Supplier Number: 4401
  • 8-Tube Strips
    1.2 ml
    12/Rack
    Sterile
    Log in Para disponibilidad
    Log in Para disponibilidad
    Case (120UN)
    Case (120UN) - $5,370.00
    $5,370.00
    Case (120UN)
       $5,370.00
    Supplier Number: 4413
  • 8-Cap Strips
    1.2 ml
    12/Bag
    Sterile
    Log in Para disponibilidad
    Log in Para disponibilidad
    Case (120UN)
    Case (120UN) - $1,430.00
    $1,430.00
    Case (120UN)
       $1,430.00
    Supplier Number: 4418