High-Strength Round-Bottom Centrifuge Tubes, Glass, Kimble Chase
Catalog # 46100-824
Supplier: DWK LIFE SCIENCES S. DE R.L. DE MX
Specifications
- Tamaño de la tapa:24–400
- Volumen:30 ml
- Descripción:Screw-Thread Tubes
- Referencia:46100-824
- RCF máximo:13100×g
- O.D.×L:24×106 mm
- Nº de proveedor:45600-30
Specifications
About this item
High strength 15 and 30 ml reusable centrifuge tubes are manufactured from Type 1, Class B, chemically strengthened borosilicate glass.
- Withstand temperatures upto 300 °C (572 °F)
- Screw-thread tubes are supplied with PTFE-faced rubber liner caps
These tubes can be centrifuged upto 13100 ×g when used with the accessory rubber adapter sleeve in a 50 ml rotor cavity.
Ordering information: Accessory sleeves and replacement caps sold separately.