Orders LinkContactUsLinkComponent
Edgeless® Mop Systems, Contec®
Edgeless® Mop Systems, Contec®
Supplier:  CONTEC, INC. MX
Certificates
Edgeless® Mop Systems, Contec®
Edgeless® Mop Systems, Contec®
4 Options Available Below
Product Details & Documents
Edgeless® Mops are made from strong, durable knitted polyester, eliminating contamination from breakage and deterioration common with cotton and rayon string mops. Cleanroom laundered Edgeless® Mops are made from a continuous tube of 100% polyester knitted fabric looped to form mop strands that will not shed or break.
  • Durable and chemical resistant
  • Continuous loop of mop strands provides improved cleanliness, eliminating cut edges
  • Textured large head band provides scrubbing power for difficult stains and debris
  • Ideal for use in sterile environments
Patented Textured and Long Textured Edgeless Mops feature unique ribbed, twisted strands that are more sorbent than traditional string mops. They have a textured head band that is ideal when additional abrasion resistance is required.
All mops are available sterilized by gamma radiation, and can be autoclaved. Validated sterile per the AAMI/ISO Guidelines to a 10-6 SAL. Certificates of sterility are available.
Stainless steel and polypropylene single- and multi-bucket systems are available. All mop system components and bucket systems are compatible with gamma, ETO, and autoclave sterilization, unless otherwise noted. Mop heads are laundered and packaged in an ISO Class 4 (FED-STD-209E Class 10/M2.5) cleanroom.
 : All mop system components are ordered separately. Mop heads work well with existing wet mop hardware, but a down-press ringer is recommended for maximum extraction. Polypropylene and stainless steel bucket systems include cart, buckets, wringer, and casters.
Product Family Options
Información del producto
  • Availability
    Price
  • 68300-353
    Edgeless® Floor Mop
    Edgeless® Floor Mop
    Log in Para disponibilidad
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1UN)
    Each (1UN) - $17,169.78
    $17,169.78
    Log in Para disponibilidad
    Case de 20 (1UN)
    Case de 20 (1UN) - $16,767.36
    $16,767.36
    Each (1UN)
       $17,169.78
    Case de 20 (1UN)
       $16,767.36
    Supplier Number: EL-MOP
  • 76270-916
    QuickConnect™ Edgeless Floor Mop Frame, PP Copolymer Frame with Stainless Steel Connector, 15.2×17.9 cm (6×7")
    QuickConnect™ Edgeless Floor Mop Frame, PP Copolymer Frame with Stainless Steel Connector, 15.2×17.9 cm (6×7")
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1UN)
    Each (1UN) - $2,967.43
    $2,967.43
    Each (1UN)
       $2,967.43
    Supplier Number: 2646F
  • 80080-578
    Edgeless® Textured Mop Head
    Edgeless® Textured Mop Head
    Log in Para disponibilidad
    Log in Para disponibilidad
    Case (30UN)
    Case (30UN) - $27,850.51
    $27,850.51
    Case (30UN)
       $27,850.51
    Supplier Number: ELMOP3
  • 68310-048
    Fiberglass Handle with Mop Head Frame, Non-Autoclavable, 162.6 cm (64") Long
    Fiberglass Handle with Mop Head Frame, Non-Autoclavable, 162.6 cm (64") Long
    Discontinued
    Supplier Number: -
  • 89073-978
    Quick-Connect Stainless Steel Mop Handle, 152.4 cm (60") Long
    Quick-Connect Stainless Steel Mop Handle, 152.4 cm (60") Long
    Discontinued
    Supplier Number: -
  • 89497-298
    Ribbed Edgeless® Mop, Polyester Tube
    Ribbed Edgeless® Mop, Polyester Tube
    Log in Para disponibilidad
    Log in Para disponibilidad
    Case (30UN)
    Case (30UN) - $22,258.24
    $22,258.24
    Case (30UN)
       $22,258.24
    Supplier Number: ELMOP5