PIG® HazMat Spill Kit in Small Wall-Mount Cabinet, New Pig
Catálogo # 56220-190
Proveedor: 2 D 2 S.A. DE C.V. MX
Especificaciones
- Capacidad:Absorbs up to 37.9 L (10 gal.)
- Referencia:56220-190
- Nº de proveedor:KIT315
Especificaciones
Acerca de este artículo
Safety-yellow, heavy-gauge, welded steel cabinet is printed "Spill Kit" for quick identification.
- Prepacked cabinet provides easy access to contents
- See-through front panel lets you easily check inventory levels
- Features an extra shelf for storing PPE
- Includes write-on label for emergency contact info
- Two adjustable shelves
Optional Refill for PIG® HazMat Spill Kit in Small Wall-Mount Cabinet is packed to refill your PIG HazMat Spill Kit. PIG HazMat Pads and Pillows absorb; Wipers clean surfaces and hands. PIG HazMat Pads and Pillows are specially treated for unsurpassed performance with concentrated corrosives, such as 98% sulfuric acid and 30% sodium hydroxide. Temporary disposal bags help make cleanup easier.
PIG HazMat Pads and Pillows absorb; wipers clean hands and surfaces. PIG HazMat Absorbents are specially treated for unsurpassed performance with concentrated corrosives, such as 98% sulfuric acid and 30% sodium hydroxide.