Sure-Grip® EX Slimline-Styled Safety Cabinets for Flammable Materials, Justrite®
Catalog # 97001-218
Supplier: JUSTRITE MANUFACTURING CO
Specifications
- Profundidad:18"
- Anchura:23¼"
- Capacidad:22 gal.
- Color:Gray
- Descripción:Safety cabinet Slimline
- Altura:65"
- Referencia:97001-218
- Peso al enviarse:211 lbs.
- Tipo:1 self-closing door, 3 shelves
- Nº de proveedor:892223
Specifications
About this item
High-capacity cabinets offer increased storage in environments with limited floor space.
- Cabinets are available in three colors, in manual or self-close formats
Double-wall, air insulated, welded steel construction provides reliable fire resistance. Single-door units are supplied with three galvanized steel shelves and a 5.1 cm (2") leakproof sump for spill control. Additional standard features include dual vents, adjustable leveling feet, a grounding connector, and a large trilingual warning label. Use alone for storage in hard-to-fit places near points of use, or alongside cabinets of the same depth for expanded storage or segregating incompatible liquids.