Orders LinkContactUsLinkComponent
VWR® Stirring Rods
VWR® Stirring Rods
PTFE with Steel Core, 6 mm, 200 mm
Catalog # 89026-280
Supplier:   VWR International
VWR® Stirring Rods
Catalog # 89026-280
Supplier:  VWR International
VWR® Stirring Rods
VWR® Stirring Rods
PTFE with Steel Core, 6 mm, 200 mm
Price
$2,281.57
Log in Para disponibilidad
Paquete de 5
Product Details & Documents
Available with a steel core or in solid PTFE, these rods are chemically resistant, inert, and non-abrasive.
  • Constructed from solid PTFE or with a steel core
  • Chemically resistant
  • Inert
  • Non-abrasive
Steel core rods have a rounded end, can be bent into a permanent shape, and are ideal when extra rigidity is required. Solid PTFE rods have a tapered end. Usable to 280°C (536°F).
Especificaciones
  • Ø:
    6 mm
  • Length:
    200 mm
  • Material:
    PTFE with Steel Core