Orders LinkContactUsLinkComponent
Temperature Probes
Temperature Probes
Temperature Probes
Certificates
Temperature Probes
Temperature Probes
2 Options Available Below
Product Details & Documents
These sensing elements are housed in glass or tubular stainless steel and are designed for connection to METTLER TOLEDO pH meters.
  • Glass or stainless steel housing
  • Lead wires isolated from outer probe
Probes have a maximum operating temperature of 150 °C (302 °F) and are constructed with the sensing elements and lead wires electrically isolated from the outer probe surface for maximum safety.
Ordering information: Probes are part of the InLab® product line and are supplied with quality certificate.
Product Family Options
Información del producto
  • Material
    Ø
    Length
    Cable Length
    Connector
    Accuracy
    Availability
    Price
  • 11278-744
    NTC 30K temperature probe
    NTC 30K temperature probe
    Stainless steel 316
    4 mm
    130 mm
    1.2 m
    RCA (cinch)
    ±0.20 °C (from –30...+100 °C)
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1UN)
    Each (1UN) - $6,641.66
    $6,641.66
    Each (1UN)
       $6,641.66
    Supplier Number: 51300164
  • 11278-746
    Pt1000 temperature probe
    Pt1000 temperature probe
    Stainless steel 316
    4 mm
    130 mm
    1.2 m
    Banana, 4 mm
    ±0.15 °C (from –30...+100 °C)
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1UN)
    Each (1UN) - $19,077.12
    $19,077.12
    Each (1UN)
       $19,077.12
    Supplier Number: 51300165