Stirring Elements, IKA® Works
Acerca de este artículo
Made of stainless steel. Accepted by overhead stirrer motors with corresponding output shaft/chuck diameter measurements.
Turbine stirrers generate axial flow, pulling material from above, with minimal shearing. Configuration reduces the risk of user injury if blade contacts the vessel wall.
Dissolver stirrers produce radial flow, drawing material from both top and bottom, with high turbulence and shearing forces for effective particle reduction.
Centrifuge stirrers have two blades that collapse for insertion into narrow-necked vessels and unfold with increasing speed. Operation is similar to four-bladed propellers.
Paddle stirrers effect tangential flow with minimal turbulence and gentle treatment of the media.
Anchor stirrers are ideal for moderately to highly viscous fluids, inducing tangential flow and high shearing rate at the perimeter and leaving minimal material deposit on the vessel wall.
Ordering information: When ordering stirring paddles, please specify the chuck size of your stirrer.
6 Options Available Below
Product Details & Documents
Made of stainless steel. Accepted by overhead stirrer motors with corresponding output shaft/chuck diameter measurements.
Turbine stirrers generate axial flow, pulling material from above, with minimal shearing. Configuration reduces the risk of user injury if blade contacts the vessel wall.
Dissolver stirrers produce radial flow, drawing material from both top and bottom, with high turbulence and shearing forces for effective particle reduction.
Centrifuge stirrers have two blades that collapse for insertion into narrow-necked vessels and unfold with increasing speed. Operation is similar to four-bladed propellers.
Paddle stirrers effect tangential flow with minimal turbulence and gentle treatment of the media.
Anchor stirrers are ideal for moderately to highly viscous fluids, inducing tangential flow and high shearing rate at the perimeter and leaving minimal material deposit on the vessel wall.
Ordering information: When ordering stirring paddles, please specify the chuck size of your stirrer.
Product Family Options
Información del producto
- ØMaximum SpeedShaft SizeAvailabilityPrice
- 33994-032R 1342 Four-Bladed Propeller StirrerR 1342 Four-Bladed Propeller Stirrer50 mm (head)2000 rpmØ×L 8×350 mmLog in Para disponibilidadEach (1UN)$13,010.79Supplier Number: 0000741000
- 33994-074R 1312 Turbine StirrerR 1312 Turbine Stirrer50 mm (head)2000 rpmØ×L 8×350 mmLog in Para disponibilidadEach (1UN)$18,933.22Supplier Number: 2333000
- 33994-116R 1382 Three-Bladed Propeller StirrerR 1382 Three-Bladed Propeller Stirrer55 mm (head)2000 rpmØ×L 8×350 mmLog in Para disponibilidadEach (1UN)$11,076.38Supplier Number: 1295900
- 89176-704R 1385 Three-Bladed Propeller StirrerR 1385 Three-Bladed Propeller Stirrer140 mm (head)800 rpm10×550 mmLog in Para disponibilidadEach (1UN)$12,151.75Supplier Number: 0477700
- 33994-124R 1300 Dissolver StirrerR 1300 Dissolver Stirrer80 mm (head)2000 rpmØ×L 8×350 mmLog in Para disponibilidadEach (1UN)$24,102.60Supplier Number: 0000513500
- 33994-138R 1330 Anchor StirrerR 1330 Anchor Stirrer45 mm (head)1000 rpmØ×L 8×550 mmLog in Para disponibilidadEach (1UN)$14,828.15Supplier Number: 2022300