Orders LinkContactUsLinkComponent
VWR® Talon® Support Stands
VWR® Talon® Support Stands
Support Stand, 451×267 mm (17.75×10.5"), 71.1 cm (28")
Catalog # 58958-459
Supplier:   TROEMNER LLC
Supplier #:  30400031
VWR® Talon® Support Stands
Catalog # 58958-459
Supplier:  TROEMNER LLC
VWR® Talon® Support Stands
VWR® Talon® Support Stands
Support Stand, 451×267 mm (17.75×10.5"), 71.1 cm (28")
Price
$12,698.25
Log in for availability
Each (1UN)
Product Details & Documents
Support stands are made entirely of Type 303 stainless steel and can accommodate vessels up to 45.7cm (18”) in diameter within the sturdy U-shaped base. The threaded stainless steel support rod is 16mm (5/8") in diameter and is screwed into the support stand base. Includes support rod.
  • Stainless steel construction
  • Sturdy U-shaped base
  • Includes support rod
  • Fitted with oversized knobs for very secure positioning
The threaded stainless steel support rod is 16 mm (5/8") in diameter and is screwed into the support stand base
Accessories include an extra stainless steel support rod, two choices of clamps, and stainless steel stabilizer bolts that are threaded to fit the extra holes in the base of the support stand. The stainless steel support stand clamp (58950-660) is designed with a precision fit to accept a 13mm (1/2") diameter support shaft and a 16mm (5/8") diameter support rod on a support stand. Heavy-duty support clamp (14010-000) is made of a strong aluminum alloy and is fitted with oversized knobs for very secure positioning. It supports all shafts from 6 to 24mm (0.24 to 0.94") in diameter and supports rods on a 10.2 cm (4") long surface to avoid wobble and vibration.
Includes support rod.
Ordering information: Assembly of support stand (base and one support rod) required. Additional support rods, clamps, and stabilizer bolts are ordered separately.
Documents
Specifications
  • Description:
    Support Stand
  • Height:
    71.1 cm (28")
  • Base Dimensions:
    451×267 mm (17.75×10.5")
Accessories
Product Information
  • Catalog #
    Rod Height
    Availability
    Price
  • Replacement Support Rod
    Replacement Support Rod
    97035-522
    58.4 cm (23")
    Log in for availability
    Each (1UN) - $3,230.33