Orders LinkContactUsLinkComponent
Clamping Systems and Support Stands, Arrow
Clamping Systems and Support Stands, Arrow
Clamping Systems and Support Stands, Arrow
Certificates
Clamping Systems and Support Stands, Arrow
Clamping Systems and Support Stands, Arrow
1 Options Available Below
Product Details & Documents
“Power Hold” Universal Clamps can be used for most applications. Designed with large knobs for powerful gripping, these clamps feature an adjustable stop for speed and safety in set up and operation. Made with precision machined aluminum rod for strength and durability. Can hold any stirrer that has a mounting rod stirring stand diameter from 9.5 – 16 mm (3/85/8") or from 12 – 19 mm (1/23/4").
Universal Swivel Clamp allows stirrer to be set at any compound angle for best stirring action
Arrow Support Stand consists of heavy duty steel base and stainless steel support rod. Base has two long front legs and one short back leg, all with rubber pads, to increase stability and cut vibration. Overall height is 73.7 cm (29"). Models available with 16 mm (5/8") or 19 mm (3/4") diameter support rod.
Spider Support Stand can be clamped on the rim of 5-gallon pails, small drums, square or rectangular tanks. Will fit securely on vessels with rims up to 16 mm (5/8") width. Stand is constructed of lightweight aluminum base and has a 61Hx1.6 dia. cm (24 x 5/8") stainless steel support rod that allows user to raise and lower stirrer. Legs and swivel clamp adjust to give maximum holding power. Models available with 5-gallon vessel or 55-gallon vessel capacity.
Note: Spider Support Stand does not rest on any kind of base and does not come with the vessel.
Product Family Options
Stands
Información del producto
  • Capacity
    Availability
    Price
  • 59025-166
    Arrow Support
    Arrow Support
    ⅝" dia. rod
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1UN)
    Each (1UN) - $10,363.83
    $10,363.83
    Each (1UN)
       $10,363.83
    Supplier Number: ASE-625