Lab Frame connector securely attaches to support stands, lab frames, or any support rod up to 1.9 cm (3/4") in diameter.
- Unique, flexible arm
- Provided with vinyl and fiberglass sleeves
- Fiberglass sleeves are suited to higher-temperature applications
- Options suited to different-shaped apparatuses
Systems are extremely versatile with a unique, flexible arm that can be placed in virtually any position or angle
The spring clamp head features locking wing nuts that apply tension to the nickel-plated jaws. It is designed to hold thermometers, glass tubes, and temperature probes.
The two- and three-prong clamps are constructed of cast alloy with dual adjust for greater flexibility. Both clamps are provided with vinyl and fiberglass sleeves. The fiberglass sleeves are recommended for higher temperature applications up to 100°C (212°F). The two-prong clamp is ideal for holding straight-sided apparatus such as burets, thermometers, beakers, and flask necks. The three-prong clamp is ideal for holding irregularly shaped objects.
Systems include a two-prong clamp, a three-prong clamp, a spring clamp head, a nickel-plated flex arm of either 30.5 cm (12") or 45.7 cm (18") length, a cast alloy Lab Frame connector, and an utility wrench.
- Catalog No:
- 12621-216
- Brazo Ø:
- 1.3 cm (1/2")