Black Phenolic Screw Caps, Rubber Liner, Qorpak®
: QORPAK
Black phenolic caps with rubber liners are autoclavable
These black phenolic screw caps are made of phenolic resin.
This liner is FDA approved for food and pharmaceutical applications. Excellent properties of resilience, resistance to moisture vapor and relatively low gas transmission. Also widely used for packaging injectables. Great for culture work. They also provide the most consistent adherence to close dimensional tolerances.
Frequently Bought Together