ContactUsLinkComponent
Green Thermoset Screw Caps, F217 and PTFE Liner, Qorpak®
Green Thermoset Screw Caps, F217 and PTFE Liner, Qorpak®
Catálogo # 75993-610
Proveedor:  QORPAK
undefined
Green Thermoset Screw Caps, F217 and PTFE Liner, Qorpak®
Catálogo # 75993-610
Proveedor:  QORPAK
Número de proveedor:  272627
Especificaciones
  • Tamaño de la tapa:
    22-400
  • Referencia:
    75993-610
Especificaciones
Acerca de este artículo
Green Thermoset Plastic Caps with F217 and PTFE liners are known for providing the widest range of chemical compatibility.
  • The color green easily identifies the F217 & PTFE liner system
  • F217 foam-backed PTFE liners resist attack from virtually all chemicals at room temperature
  • Provides odor and impact resilience for a tight seal with a low moisture transmission rate
The F217 features a low-density foam core between two LDPE discs. They are applied to the back of a PTFE liner.
Some uses for these F217/PTFE lined caps include analytical lab samples, high purity chemicals, strong acids solvents, environmental samples, pharmaceuticals, and diagnostic reagents.