NALGENE® Polypropylene Filling/Venting Closures, Thermo Scientific
Supplier: Thermo Fisher Scientific
Certificates
About this item
Polypropylene closures for aseptic liquid transfer of media, biological reagents, pure water, and chemicals
Frequently used to feed or receive from bioreactors and fermentors. Can be used with carboys or bottles that accept 53B or 83B closures. Filling/Venting Closures are provided with two pieces of tissue culture-grade tubing for aspiration, dispensing, and filling splash guard. The TopWorks closure includes two 6.4mm (1/4") rubber tubes and one 9.5mm (3/8") rubber tube cured to and passing through a silicone rubber stopper/liner in an 83B closure. It is compliant with USP Class VI requirements. All closures are autoclavable.
Related Information
Frequently Bought Together