VWR® Centrifugal Filters
Catalog # 82031-360
Supplier: VWR International
Product available on GSA Advantage®
Specifications
- Tamaño de poro:0.45 µm
- Largo:4.5 cm (125/32")
- Color:Berry
- Descripción:Nylon membrane
- Rango de temperatura de funcionamiento:0...40 °C (32...104 °F)
- Max. sample volume:500 µl
- Volumen de retención:5 µl
- Área efectiva de filtrado:0.28 cm²
- Max. centrifugal force:14000 g
- Max. receiver volume:500 µl
- Referencia:82031-360
- MWCO:—
Specifications
About this item
Use to: Concentrate, purify, and desalt peptides, proteins, oligonucleotides, DNA, and RNA; clean up labeling and PCR reactions; isolate DNA from agarose gel slices; buffer exchange; separate proteins, oligonucleotides, and RNA from acrylamide gels; clarify samples; or remove particulates.
- Polypropylene housing
- Membrane options available
Filter housings are constructed of polypropylene and are available with a polyethersulfone (PES) or nylon membrane.
Filters with nylon membrane are ideal for prefiltration clean-up of large agarose or acrylamide gel debris prior to further purification with an MWCO device.