About this item
Specially designed for automatic sample preparation for the clarification of samples. Filters provide fast and efficient filtration of aqeuous and organic solutions. Housing is constructed of mechanically stable polypropylene. External geometry of filter housing ensures smooth filter transport from storage turntable to filtration site, and easy filter changing. Female Luer-lock inlet for secure fit to syringe barrel and male Luer slip outlet. Diameter: 25mm.
- Optimized for SOTAX™, Caliper (Zymark™), Varian™, and Erweka™ tablet tester systems
- Mechanically stable polypropylene housing specifically designed for automated filtration
- Recyclable after use
Automated filtration using an autosampler greatly reduces the time and difficulty of sample preparation for tablet dissolution testing in a high-throughput laboratory.
Roby syringe filters have a polypropylene housing specifically designed for use in commonly available tablet tester systems to support this type of automated filtration.
Related Information
Certificates
Search by Lot NumberCytiva CyDye Licensing Statements Cytiva General Licensing Statements