Orders LinkContactUsLinkComponent
Whatman™ Polycap™ TC Disposable Filter Capsules
Whatman™ Polycap™ TC Disposable Filter Capsules
 :   Cytiva
Whatman™ Polycap™ TC Disposable Filter Capsules
 :  Cytiva
Whatman™ Polycap™ TC Disposable Filter Capsules
Whatman™ Polycap™ TC Disposable Filter Capsules
Whatman Polycap TC filtration capsules from Cytiva feature dual layer polyethersulfone (PES) membranes and provide efficient filtration for aqueous solutions.
  • 100% integrity-tested during manufacturing
  • Polypropylene housing thermally fused to eliminate surfactants and mold releasing agents that could affect analysis
  • Available in sterile and nonsterile versions with a filling bell option
  • PES membranes protein adsorption characteristics: HAS 0.4 µg/cm²; Insulin 2.0 µg/cm²; gammaglobulin 1.5 µg/cm²
PES membranes are inherently hydrophilic, have low extractables, are biosafe, and have excellent flow rates. These characteristics make Polycap TC PES filters appropriate for a variety of critical filtration applications, including cleaning/rinsing pharmaceutical solutions, tissue culture media, and viral suspensions.
Certifications: Manufactured according to ISO9001:2008 QMS standards.
  • Catalog No:
  • 30253-228
  • 83009-778
  • 30253-244
  • 83009-780
  • 30253-260
  • 83009-782
  • Autoclavable:
  • Certain filter devices have been sterilized.
    Capsule may be autoclaved at 121 °C for 20 minutes. However, an integrity test should be performed after autoclaving.
  • Certain filter devices have been sterilized.
    Capsule may be autoclaved at 121 °C for 20 minutes. However, an integrity test should be performed after autoclaving.
  • Certain filter devices have been sterilized.
    Capsule may be autoclaved at 121 °C for 20 minutes. However, an integrity test should be performed after autoclaving.
  • Certain filter devices have been sterilized.
    Capsule may be autoclaved at 121 °C for 20 minutes. However, an integrity test should be performed after autoclaving.
  • Certain filter devices have been sterilized.
    Capsule may be autoclaved at 121 °C for 20 minutes. However, an integrity test should be performed after autoclaving.
  • Certain filter devices have been sterilized.
    Capsule may be autoclaved at 121 °C for 20 minutes. However, an integrity test should be performed after autoclaving.
  • Endotoxin level:
  • LAL tested, ≤0.5 EU/ml
  • LAL tested, ≤0.5 EU/ml
  • LAL tested, ≤0.5 EU/ml
  • LAL tested, ≤0.5 EU/ml
  • LAL tested, ≤0.5 EU/ml
  • LAL tested, ≤0.5 EU/ml
  • Flow direction:
  • If there is a prefilter, it is located on the inlet side and flow should follow arrows.
  • If there is a prefilter, it is located on the inlet side and flow should follow arrows.
  • If there is a prefilter, it is located on the inlet side and flow should follow arrows.
  • If there is a prefilter, it is located on the inlet side and flow should follow arrows.
  • If there is a prefilter, it is located on the inlet side and flow should follow arrows.
  • If there is a prefilter, it is located on the inlet side and flow should follow arrows.
  • Max. operating pressure:
  • 4.1 bar (60 psi)
  • 4.1 bar (60 psi)
  • 4.1 bar (60 psi)
  • 4.1 bar (60 psi)
  • 4.1 bar (60 psi)
  • 4.1 bar (60 psi)
  • Membrane:
  • Polyethersulfone (PES)
  • Polyethersulfone (PES)
  • Polyethersulfone (PES)
  • Polyethersulfone (PES)
  • Polyethersulfone (PES)
  • Polyethersulfone (PES)
  • Sealing:
  • Heat-fused
  • Heat-fused
  • Heat-fused
  • Heat-fused
  • Heat-fused
  • Heat-fused
  • Vent:
  • On inlet
  • On inlet
  • On inlet
  • On inlet
  • On inlet
  • On inlet
Product Family Options
  • Description
    Pore size
    Inlet/outlet Connections
    Sterility
  • Polycap TC 36 capsule filter, plus filling bell
    0.2/0.2 µm
    Stepped barb for 6 - 10 mm (¹/₄ - ³/₈") tubing
    Sterile
    Each (1UN)
    Each (1UN) - $4,710.80
    $4,710.80
    Each (1UN)
       $4,710.80
     : 6715-3602
    Product available on GSA Advantage®
    Specifications
    Cat. No.
    30253-228
    Autoclavable
    Certain filter devices have been sterilized.
    Capsule may be autoclaved at 121 °C for 20 minutes. However, an integrity test should be performed after autoclaving.
    Endotoxin level
    LAL tested, ≤0.5 EU/ml
    Flow direction
    If there is a prefilter, it is located on the inlet side and flow should follow arrows.
    Max. operating pressure
    4.1 bar (60 psi)
    Membrane
    Polyethersulfone (PES)
    Sealing
    Heat-fused
    Vent
    On inlet
    Product available on GSA Advantage®
  • Polycap TC 36 capsule filter, plus filling bell
    0.8/0.2 µm
    Stepped barb for 6 - 10 mm (¹/₄ - ³/₈") tubing
    Sterile
    Each (1UN)
    Each (1UN) - $4,158.82
    $4,158.82
    Each (1UN)
       $4,158.82
     : 6715-3682
    Product available on GSA Advantage®
    Specifications
    Cat. No.
    83009-778
    Autoclavable
    Certain filter devices have been sterilized.
    Capsule may be autoclaved at 121 °C for 20 minutes. However, an integrity test should be performed after autoclaving.
    Endotoxin level
    LAL tested, ≤0.5 EU/ml
    Flow direction
    If there is a prefilter, it is located on the inlet side and flow should follow arrows.
    Max. operating pressure
    4.1 bar (60 psi)
    Membrane
    Polyethersulfone (PES)
    Sealing
    Heat-fused
    Vent
    On inlet
    Product available on GSA Advantage®
  • Polycap TC 75 capsule filter, plus filling bell
    0.2/0.2 µm
    Stepped barb for 6 - 10 mm (¹/₄ - ³/₈") tubing
    Sterile
    Each (1UN)
    Each (1UN) - $7,231.18
    $7,231.18
    Each (1UN)
       $7,231.18
     : 6715-7502
    Product available on GSA Advantage®
    Specifications
    Cat. No.
    30253-244
    Autoclavable
    Certain filter devices have been sterilized.
    Capsule may be autoclaved at 121 °C for 20 minutes. However, an integrity test should be performed after autoclaving.
    Endotoxin level
    LAL tested, ≤0.5 EU/ml
    Flow direction
    If there is a prefilter, it is located on the inlet side and flow should follow arrows.
    Max. operating pressure
    4.1 bar (60 psi)
    Membrane
    Polyethersulfone (PES)
    Sealing
    Heat-fused
    Vent
    On inlet
    Product available on GSA Advantage®
  • Polycap TC 75 capsule filter, plus filling bell
    0.8/0.2 µm
    Stepped barb for 6 - 10 mm (¹/₄ - ³/₈") tubing
    Sterile
    Each (1UN)
    Each (1UN) - $6,390.08
    $6,390.08
    Each (1UN)
       $6,390.08
     : 6715-7582
    Product available on GSA Advantage®
    Specifications
    Cat. No.
    83009-780
    Autoclavable
    Certain filter devices have been sterilized.
    Capsule may be autoclaved at 121 °C for 20 minutes. However, an integrity test should be performed after autoclaving.
    Endotoxin level
    LAL tested, ≤0.5 EU/ml
    Flow direction
    If there is a prefilter, it is located on the inlet side and flow should follow arrows.
    Max. operating pressure
    4.1 bar (60 psi)
    Membrane
    Polyethersulfone (PES)
    Sealing
    Heat-fused
    Vent
    On inlet
    Product available on GSA Advantage®
  • Polycap TC 150 capsule filter, plus filling bell
    0.2/0.2 µm
    Stepped barb for 10 - 12 mm (³/₈ - ¹/₂") tubing
    Sterile
    Each (1UN)
    Each (1UN) - $9,479.21
    $9,479.21
    Each (1UN)
       $9,479.21
     : 6718-9502
    Product available on GSA Advantage®
    Specifications
    Cat. No.
    30253-260
    Autoclavable
    Certain filter devices have been sterilized.
    Capsule may be autoclaved at 121 °C for 20 minutes. However, an integrity test should be performed after autoclaving.
    Endotoxin level
    LAL tested, ≤0.5 EU/ml
    Flow direction
    If there is a prefilter, it is located on the inlet side and flow should follow arrows.
    Max. operating pressure
    4.1 bar (60 psi)
    Membrane
    Polyethersulfone (PES)
    Sealing
    Heat-fused
    Vent
    On inlet
    Product available on GSA Advantage®
  • Polycap TC 150 capsule filter, plus filling bell
    0.8/0.2 µm
    Stepped barb for 10 - 12 mm (³/₈ - ¹/₂") tubing/Stepped barb for 6 - 10 mm (¹/₄ - ³/₈") tubing
    Sterile
    Each (1UN)
    Each (1UN) - $7,006.60
    $7,006.60
    Each (1UN)
       $7,006.60
     : 6718-9582
    Specifications
    Cat. No.
    83009-782
    Autoclavable
    Certain filter devices have been sterilized.
    Capsule may be autoclaved at 121 °C for 20 minutes. However, an integrity test should be performed after autoclaving.
    Endotoxin level
    LAL tested, ≤0.5 EU/ml
    Flow direction
    If there is a prefilter, it is located on the inlet side and flow should follow arrows.
    Max. operating pressure
    4.1 bar (60 psi)
    Membrane
    Polyethersulfone (PES)
    Sealing
    Heat-fused
    Vent
    On inlet