Pleated Capsule Filters, Cytiva (Formerly Pall Lab)
Catalog # 28145-757
Supplier: Cytiva
Product available on GSA Advantage®
Specifications
- Tamaño de poro:0.2 µm
- Descripción:Vented, Sterile
- Referencia:28145-757
- Volumen de elución:1350 cm2
- Nº de proveedor:12117
Specifications
About this item
Designed both for prefiltration of particulate liquids (such as serum) prior to sterilizing filtration, and removal of debris and particles from liquids not requiring sterilization.
- Maximum operating temperature: 88 °C (190 °F)
- Maximum operating pressure: 3.4 bar (50 psi)
- Polycarbonate housing
- Vented capsules
Pleated Versapor® supported acrylic copolymer membrane filter in a polycarbonate housing
Both capsules are vented and have 1.27 cm (¹/₂") hose barb inlet/outlet connections. Overall length: 21.7 cm (8¹/₂"). The presterilized filter has a hold-up volume of 50 ml, and the nonsterile filter has a hold-up volume of 42 ml.
Packaging: Individually packed, ready-to-use, and disposable.