Orders LinkContactUsLinkComponent
VWR® Tissue Grinders, PTFE
VWR® Tissue Grinders, PTFE
Supplier:   VWR International
VWR® Tissue Grinders, PTFE
Supplier:  VWR International
Certificates
VWR® Tissue Grinders, PTFE
VWR® Tissue Grinders, PTFE
3 Options Available Below
Product Details & Documents
Composed of a stainless steel plunger with pure PTFE head and a precision borosilicate glass vessel.
  • Inert
  • Non-contaminating
  • PTFE head
  • Available in plain or serrated formats
Plungers are available plain for softer tissues, or serrated for harder tissues. Tissue grinders are inert and non-contaminating. Standard clearance (between plunger and glass): 0.15–0.25mm.
Ordering information: Glass vessel and plunger are sold separately. Please order corresponding sizes of each.
Product Family Options
Glass vessels
Información del producto
  • Capacity
    Ø
    Height
    Availability
    Price
  • 89026-386
    Glass Vessel
    Glass Vessel
    2 ml
    8 mm
    120 mm
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1UN)
    Each (1UN) - $2,089.40
    $2,089.40
    Each (1UN)
       $2,089.40
  • 89026-382
    Glass Vessel
    Glass Vessel
    10 ml
    15 mm
    150 mm
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1UN)
    Each (1UN) - $2,588.29
    $2,588.29
    Each (1UN)
       $2,588.29
Plain plungers
Información del producto
  • Capacity
    Ø
    Height
    Availability
    Price
  • 89026-394
    Plain Plunger
    Plain Plunger
    10 ml
    270 mm
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1UN)
    Each (1UN) - $918.89
    $918.89
    Each (1UN)
       $918.89