ContactUsLinkComponent
Whatman™ Polydisc™ TF In-Line Filters
Whatman™ Polydisc™ TF In-Line Filters
Proveedor:  Cytiva
Certificados
undefined
Whatman™ Polydisc™ TF In-Line Filters
Proveedor:  Cytiva
Certificados
Especificaciones
  • Catalog No:
  • 28137-852
  • 28137-856
  • Aplicación:
  • Pharmaceutical: vents and in-line applications.
    Biotech: sterile vents and exhausts for growth environments, in-line sterilization of gases.
    Laboratory: clean or sterile gases. Filtration of solvents and reagents. Drying gases.
    Electronics: photoresists, solvents, gases for research.
  • Pharmaceutical: vents and in-line applications.
    Biotech: sterile vents and exhausts for growth environments, in-line sterilization of gases.
    Laboratory: clean or sterile gases. Filtration of solvents and reagents. Drying gases.
    Electronics: photoresists, solvents, gases for research.
  • Volumen de llenado:
  • 540 µl
  • 540 µl
  • Housing material:
  • Polypropylene (PP)
  • Polypropylene (PP)
  • Membrane:
  • Polytetrafluoroethylene (PTFE)
  • Polytetrafluoroethylene (PTFE)