VWR® Redi-Stor™ Conductivity Probe Storage Solution
454 g (16 oz.)
Supplier: VWR International
VWR® Redi-Stor™ Conductivity Probe Storage Solution
Catalog # 62344-940
Supplier: VWR International
CAS Number:
Jump to:
Product Details & Documents
Ideal solution for storing conductivity probes.
- No conditioning required after removal from storage
- Keeps probe clean
Probe stays clean and ready for immediate use with no conditioning necessary. Eliminates growths common when storing in only water.
Documents
Especificaciones
Especificaciones
- Size:454 g (16 oz.)