Whatman™ Grade GF/F Glass Microfiber Filters, Binder Free
Supplier: Cytiva
Certificates
About this item
These borosilicate glass filters are ideal for measuring landfill leachate composition with a TCLP test. They can also be used for DNA binding and purification, filtering finely precipitated proteins and filtering biochemical solutions, fluids, and nucleic acids.
- Very fine particle retention
- Good loading capacity and flow rate
- Binder free
Specifications
- Catalog No:
- 28497-905
- 28497-907
- 28497-925
- 28497-958
- 28497-973
- Velocidad de filtración:
- Medium; 325 s (Herzberg)
- Medium; 325 s (Herzberg)
- Medium; 325 s (Herzberg)
- Medium; 325 s (Herzberg)
- Medium; 325 s (Herzberg)
- Flujo de aire:
- 19 s/100 ml/in²
- 19 s/100 ml/in²
- 19 s/100 ml/in²
- 19 s/100 ml/in²
- 19 s/100 ml/in²
- Peso base:
- 75 g/m²
- 75 g/m²
- 75 g/m²
- 75 g/m²
- 75 g/m²
- Temperatura máximo de operación:
- 550 °C
- 550 °C
- 550 °C
- 550 °C
- 550 °C
- Retención de partículas en líquido:
- 0.7 µm
- 0.7 µm
- 0.7 µm
- 0.7 µm
- 0.7 µm
- Espesor típico:
- 420 µm
- 420 µm
- 420 µm
- 420 µm
- 420 µm
- Caudal de agua típica:
- 41 ml/min
- 41 ml/min
- 41 ml/min
- 41 ml/min
- 41 ml/min
Specifications
Related Information
Certificates
Search by Lot NumberCytiva CyDye Licensing Statements Cytiva General Licensing Statements
Additional Product Details
Frequently Bought Together