VWR® Qualitative Crepe Filter Paper, Grade 417
Good for rapid filtrations where fine particle retention is not required.
- Coarse porosity
- Fast flow rate
- Assorted sizes
- Catalog No:
- 28313-181
- 28313-057
- 28313-046
- 28313-068
- Retención de partículas:
- 40 µm
- 40 µm
- 40 µm
- 40 µm
- Porosidad:
- Coarse
- Coarse
- Coarse
- Coarse
Frequently Bought Together