Orders LinkContactUsLinkComponent
Whatman™ Quantitative Filter Papers, Hardened Ashless Grades, Grade 541
Whatman™ Quantitative Filter Papers, Hardened Ashless Grades, Grade 541
Supplier:   Cytiva
Whatman™ Quantitative Filter Papers, Hardened Ashless Grades, Grade 541
Supplier:  Cytiva
Certificates
Whatman™ Quantitative Filter Papers, Hardened Ashless Grades, Grade 541
Whatman™ Quantitative Filter Papers, Hardened Ashless Grades, Grade 541
6 Options Available Below
Product Details & Documents
Cytiva's Whatman Grade 541 acid hardened cellulose filter paper features high wet-strength, high chemical resistance, and a large nominal particle retention for fast analytical filtration.
  • High wet-strength
  • High chemical resistance to strong acid and alkali suspensions
  • Retention of gelatinous precipitate
  • 22 µm nominal particle retention rating
  • Fast flow rate (Herzberg filtration speed 34 s)
  • Maximum of 0.006% ash concentration
Along with fast Grade 541 filter paper, the hardened, ashless range from Cytiva's business includes general-purpose Grade 540 and Grade 542 for fine particle retention. All three paper grades are appropriate for use in gravimetric analysis with strong acids and alkali samples.
Hardened ashless filter paper is recommended when fast filtration of a gelatinous precipitate for gravimetric analysis is performed on a strongly acidic or alkali solution.
Applications of Grade 541 papers include:
- Use as an animal foodstuff filter for determining fiber content
- Determination of gelatin content in milk and cream
- Quantification of chloride in cement, as well as chloride and phosphorous in coal and coke
Specifications for Product Family
Especificaciones
  • Catalog No:
  • 28479-016
  • 28479-027
  • 28479-060
  • 28479-082
  • 28479-129
  • 28479-130
  • Grade:
  • Grade 541
  • Grade 541
  • Grade 541
  • Grade 541
  • Grade 541
  • Grade 541
  • Thickness:
  • 155 µm (typical)
  • 155 µm (typical)
  • 155 µm (typical)
  • 155 µm (typical)
  • 155 µm (typical)
  • 155 µm (typical)
  • Filtration speed:
  • 34 s (Herzberg)
  • 34 s (Herzberg)
  • 34 s (Herzberg)
  • 34 s (Herzberg)
  • 34 s (Herzberg)
  • 34 s (Herzberg)
  • Airflow rate:
  • 3 s/100 ml/sq. in.
  • 3 s/100 ml/sq. in.
  • 3 s/100 ml/sq. in.
  • 3 s/100 ml/sq. in.
  • 3 s/100 ml/sq. in.
  • 3 s/100 ml/sq. in.
  • Basis weight:
  • 82 g/m²
  • 82 g/m²
  • 82 g/m²
  • 82 g/m²
  • 82 g/m²
  • 82 g/m²
  • Burst strength:
  • 0.365 bar, wet burst; 0.965 bar, dry burst
  • 0.365 bar, wet burst; 0.965 bar, dry burst
  • 0.365 bar, wet burst; 0.965 bar, dry burst
  • 0.365 bar, wet burst; 0.965 bar, dry burst
  • 0.365 bar, wet burst; 0.965 bar, dry burst
  • 0.365 bar, wet burst; 0.965 bar, dry burst
  • Flow rate:
  • Fast
  • Fast
  • Fast
  • Fast
  • Fast
  • Fast
  • Tensile strength:
  • 43.4 N/15 mm Tensile M/D Dry
  • 43.4 N/15 mm Tensile M/D Dry
  • 43.4 N/15 mm Tensile M/D Dry
  • 43.4 N/15 mm Tensile M/D Dry
  • 43.4 N/15 mm Tensile M/D Dry
  • 43.4 N/15 mm Tensile M/D Dry
Especificaciones
Product Family Options
Información del producto
  • Material
    Dimensions
    Type
    Availability
    Price
  • 28479-016
    Grade 541 ashless fast filter paper, circles, 100 pieces
    Grade 541 ashless fast filter paper, circles, 100 pieces
    High-quality cotton linters
    55 mm
    Hardened ashless
    Log in Para disponibilidad
    Log in Para disponibilidad
    Paquete de 100
    Paquete de 100 - $1,343.86
    $1,343.86
    Paquete de 100
       $1,343.86
    Supplier Number: 1541-055
    Especificaciones:
    Product available on GSA Advantage®
    More
    Specifications
    Cat. No.
    28479-016
    Grade
    Grade 541
    Thickness
    155 µm (typical)
    Filtration speed
    34 s (Herzberg)
    Airflow rate
    3 s/100 ml/sq. in.
    Basis weight
    82 g/m²
    Burst strength
    0.365 bar, wet burst; 0.965 bar, dry burst
    Flow rate
    Fast
    Tensile strength
    43.4 N/15 mm Tensile M/D Dry
    Product available on GSA Advantage®
  • 28479-027
    Grade 541 ashless fast filter paper, circles, 100 pieces
    Grade 541 ashless fast filter paper, circles, 100 pieces
    High-quality cotton linters
    70 mm
    Hardened ashless
    Log in Para disponibilidad
    Log in Para disponibilidad
    Paquete de 100
    Paquete de 100 - $1,588.13
    $1,588.13
    Paquete de 100
       $1,588.13
    Supplier Number: 1541-070
    Especificaciones:
    Product available on GSA Advantage®
    More
    Specifications
    Cat. No.
    28479-027
    Grade
    Grade 541
    Thickness
    155 µm (typical)
    Filtration speed
    34 s (Herzberg)
    Airflow rate
    3 s/100 ml/sq. in.
    Basis weight
    82 g/m²
    Burst strength
    0.365 bar, wet burst; 0.965 bar, dry burst
    Flow rate
    Fast
    Tensile strength
    43.4 N/15 mm Tensile M/D Dry
    Product available on GSA Advantage®
  • 28479-060
    Grade 541 ashless fast filter paper, circles, 100 pieces
    Grade 541 ashless fast filter paper, circles, 100 pieces
    High-quality cotton linters
    110 mm
    Hardened ashless
    Log in Para disponibilidad
    Log in Para disponibilidad
    Paquete de 100
    Paquete de 100 - $2,409.90
    $2,409.90
    Paquete de 100
       $2,409.90
    Supplier Number: 1541-110
    Especificaciones:
    Product available on GSA Advantage®
    More
    Specifications
    Cat. No.
    28479-060
    Grade
    Grade 541
    Thickness
    155 µm (typical)
    Filtration speed
    34 s (Herzberg)
    Airflow rate
    3 s/100 ml/sq. in.
    Basis weight
    82 g/m²
    Burst strength
    0.365 bar, wet burst; 0.965 bar, dry burst
    Flow rate
    Fast
    Tensile strength
    43.4 N/15 mm Tensile M/D Dry
    Product available on GSA Advantage®
  • 28479-082
    Grade 541 ashless fast filter paper, circles, 100 pieces
    Grade 541 ashless fast filter paper, circles, 100 pieces
    High-quality cotton linters
    125 mm
    Hardened ashless
    Log in Para disponibilidad
    Log in Para disponibilidad
    Paquete de 100
    Paquete de 100 - $2,642.49
    $2,642.49
    Paquete de 100
       $2,642.49
    Supplier Number: 1541-125
    Especificaciones:
    Product available on GSA Advantage®
    More
    Specifications
    Cat. No.
    28479-082
    Grade
    Grade 541
    Thickness
    155 µm (typical)
    Filtration speed
    34 s (Herzberg)
    Airflow rate
    3 s/100 ml/sq. in.
    Basis weight
    82 g/m²
    Burst strength
    0.365 bar, wet burst; 0.965 bar, dry burst
    Flow rate
    Fast
    Tensile strength
    43.4 N/15 mm Tensile M/D Dry
    Product available on GSA Advantage®
  • 28479-129
    Grade 541 ashless fast filter paper, circles, 100 pieces
    Grade 541 ashless fast filter paper, circles, 100 pieces
    High-quality cotton linters
    185 mm
    Hardened ashless
    Log in Para disponibilidad
    Log in Para disponibilidad
    Paquete de 100
    Paquete de 100 - $6,096.96
    $6,096.96
    Paquete de 100
       $6,096.96
    Supplier Number: 1541-185
    Especificaciones:
    Product available on GSA Advantage®
    More
    Specifications
    Cat. No.
    28479-129
    Grade
    Grade 541
    Thickness
    155 µm (typical)
    Filtration speed
    34 s (Herzberg)
    Airflow rate
    3 s/100 ml/sq. in.
    Basis weight
    82 g/m²
    Burst strength
    0.365 bar, wet burst; 0.965 bar, dry burst
    Flow rate
    Fast
    Tensile strength
    43.4 N/15 mm Tensile M/D Dry
    Product available on GSA Advantage®
  • 28479-130
    Grade 541 ashless fast filter paper, circles, 100 pieces
    Grade 541 ashless fast filter paper, circles, 100 pieces
    High-quality cotton linters
    240 mm
    Hardened ashless
    Log in Para disponibilidad
    Log in Para disponibilidad
    Paquete de 100
    Paquete de 100 - $9,625.05
    $9,625.05
    Paquete de 100
       $9,625.05
    Supplier Number: 1541-240
    Especificaciones:
    Product available on GSA Advantage®
    More
    Specifications
    Cat. No.
    28479-130
    Grade
    Grade 541
    Thickness
    155 µm (typical)
    Filtration speed
    34 s (Herzberg)
    Airflow rate
    3 s/100 ml/sq. in.
    Basis weight
    82 g/m²
    Burst strength
    0.365 bar, wet burst; 0.965 bar, dry burst
    Flow rate
    Fast
    Tensile strength
    43.4 N/15 mm Tensile M/D Dry
    Product available on GSA Advantage®