ContactUsLinkComponent
Whatman™ Multiple Vacuum Filtration Apparatus, Three and Six-Place Vacuum Filtration System, Whatman products (Cytiva)
Whatman™ Multiple Vacuum Filtration Apparatus, Three and Six-Place Vacuum Filtration System, Whatman products (Cytiva)
Catalog # 97048-734
Supplier:  Cytiva
Product available on GSA Advantage®
undefined
Whatman™ Multiple Vacuum Filtration Apparatus, Three and Six-Place Vacuum Filtration System, Whatman products (Cytiva)
Catalog # 97048-734
Supplier:  Cytiva
Supplier Number:  10498762
Product available on GSA Advantage®
Specifications
  • Descripción:
    Six-place vacuum filtration system, stainless steel filter funnel (recommended for microbiology monitors and analytical funnels)
  • Referencia:
    97048-734
  • Nº de proveedor:
    10498762
Specifications
About this item
Multiple vacuum filtration apparatus suitable only for vacuum operation.
  • For microbiological quality control
  • Residue analyses
  • Serial filtration carried out rapidly and easily with a common drainage outlet
Stainless steel including sieve filter support
Individual stopcocks, 47/50 mm filter size, tubing nozzle 9 mm vacuum connection.
Caution: Suitable only for vacuum. Do not exceed 1.3bar (300mbar overpressure) when using flushing tubes.