Orders LinkContactUsLinkComponent
Canopy Hood, Labconco®
30", 18", 48"
Catalog # 30192-104
Supplier:   Labconco
 :  4799000
Canopy Hood, Labconco®
Catalog # 30192-104
Supplier:  Labconco
Canopy Hood, Labconco®
Canopy Hood, Labconco®
30", 18", 48"
Price
$74,010.00
Log in Para disponibilidad
Each (1UN)
Product Details & Documents
Specifically designed to solve unique ventilation problems, this canopy hood vents nontoxic materials such as heat, steam, and odors from large or bulky equipment. The epoxy-coated steel canopy has a built-in baffle to increase air velocities and enhance its overall capture efficiency. The canopy may be installed on a wall or suspended from the ceiling* for peninsula and corner locations. For most efficient performance, the canopy hood should be installed up to 30.5cm (12") above the equipment to be ventilated.
*For ceiling mount, threaded rods (not included) may be required.
*For ceiling mount, threaded rods (not included) may be required.
The epoxy-coated steel canopy has a built-in baffle to increase air velocities and enhance its overall capture efficiency. The canopy may be installed on a wall or suspended from the ceiling* for peninsula and corner locations. For most efficient performance, the canopy hood should be installed up to 30.5 cm (12") above the equipment to be ventilated.
Especificaciones
  • Height:
    18"
  • Depth:
    30"
  • Width:
    48"