Orders LinkContactUsLinkComponent
Wash Rings, Labconco
Wash Rings, Labconco
Wash Rings, Labconco
Certificates
Wash Rings, Labconco
Wash Rings, Labconco
1 Options Available Below
Product Details & Documents
Wash rings are suitable for use in perchloric acid duct systems.
Each ring features a wide-angle conical spray nozzle and wash water connector nipple fabricated into a PVC coupling. Use these rings with a 25.4 cm (10") or 30.5 cm (12") nominal duct.
Documents
Product Family Options
Información del producto
  • For Use With
    Availability
    Price
  • 82006-966
    Wash Rings
    Wash Rings
    25.4 cm (10") nominal duct
    Discontinued
    Supplier Number: 4746000
  • 82006-968
    Wash Rings
    Wash Rings
    30.5cm (12") nominal duct
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1UN)
    Each (1UN) - $14,140.00
    $14,140.00
    Each (1UN)
       $14,140.00
    Supplier Number: 4746100