ContactUsLinkComponent
Purospher™ STAR HPLC Columns and Cartridges, EMD Millipore
Purospher™ STAR HPLC Columns and Cartridges, EMD Millipore
Catálogo # 48219-482
undefined
Purospher™ STAR HPLC Columns and Cartridges, EMD Millipore
Catálogo # 48219-482
Número de proveedor:  1502520001
Especificaciones
  • Formato de columna:
    Cartridge
  • Ø int.:
    4.00 mm
  • Tamaño de partícula:
    5 µm
  • Tamaño de poro:
    120 Å
  • Descripción:
    LiChroCART® Stainless Steel Purospher™ STAR Cartridges
  • Longitud:
    250 mm
  • Environmentally Preferable:
  • Rango de pH:
    1.5 to 10.5
  • Referencia:
    48219-482
  • Packing Material:
    Silica
  • Phase:
    RP-18e
  • Nº de proveedor:
    1.50252.0001
Especificaciones
Acerca de este artículo
Columns are for use with basic, neutral, metal, chelating, or any other sample format, and are ideal for the separation of difficult hydrophobic samples. Purospher™ STAR columns are designed to provide optimum results even when loaded with large or small sample volumes and are also available in a narrow-bore format (1mm). The wide pH range provides the required pH stability for 99% of the common analysis applications. These columns provide the power to separate even the most difficult of compound mixtures. With up to 150,000 plates per meter for the 3µm particle size and 100,000 plates per meter for the 5µm particle size, Purospher™ STAR columns ensure good baseline separations. Special derivatization and endcapping processes ensure long-term chemical stability, excellent retention stability, and long column lifetime. All Purospher™ STAR columns are guaranteed to provide absolutely reproducible results.
Purospher™ STAR columns are designed to provide optimum results even when loaded with large or small sample volumes and are also available in a narrow-bore format (1 mm). The wide pH range provides the required pH stability for 99% of the common analysis applications. These columns provide the power to separate even the most difficult of compound mixtures. With up to 150000 plates per meter for the 3 µm particle size and 100000 plates per meter for the 5 µm particle size, Purospher™ STAR columns ensure good baseline separations. Special derivatization and endcapping processes ensure long-term chemical stability, excellent retention stability, and long column lifetime. All Purospher™ STAR columns are guaranteed to provide absolutely reproducible results.
Información de pedido: All LiChroCART® cartridges require end fittings. manuCART® and LiChroCART® series end fittings and holders are available separately. Hibar® columns include end fittings. If 4×4 mm guard columns are to be used on 4 or 4.6 mm Int.Ø. Hibar® Purospher™ STAR columns, the top fitting must be replaced with guard column holder designed to accommodate this size guard column. Other larger Int.Ø. Purospher™ STAR columns include end fittings as indicated in their descriptions. Guard columns for these larger Int.Ø columns are also available, contact your Avantor sales representative to order.