Steri-Wrap® I, Propper Manufacturing
Steri-Wrap® I, 30.5W x 30.5L cm (12 x 12")
Supplier: PROPPER MFG CO INC
: 31101200
Jump to:
Product Details & Documents
Steri-Wrap® I is a medium weight wrap, ideal for general hospital use in steam and ethylene oxide sterilization cycles.
- Cellulose based wrap for Steam and EO sterilization
- Efficient microbial barrier, high-level of protection against fluids and particles
- Highly permeable meaning no wet packs
- Variety of strength of fabric to meet your needs
Steri-Wrap® I is cellulose based sterilization wrap available in three grades for keeping packages safe and secure during the sterilization process. Steri-Wrap® I is a medium weight wrap, ideal for general hospital use. Steri-Wrap® I is designed to be virtually lint free with tightly anchored fibers. Wrap is reversible and features excellent liquid repellency.
Certifications: ISO 11607-1:2019
Documents
Especificaciones
- Description:Steri-Wrap® I
- Dimensions:30.5W x 30.5L cm (12 x 12")