Orders LinkContactUsLinkComponent
EasyDip™ Slide Staining System, Simport Scientific
EasyDip™ Slide Staining System, Simport Scientific
EasyDip™ Slide Staining System, Simport Scientific
Certificates
EasyDip™ Slide Staining System, Simport Scientific
EasyDip™ Slide Staining System, Simport Scientific
2 Options Available Below
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
The EasyDip™ slide staining system consists of a square staining jar and a 12-position vertical slide rack. Ideal for special stains, frozen sections, and microwave oven processes, the system is easy to clean and autoclavable. The slide staining jars have a wide, stable base for greater stability while the recessed interior allows for a smaller reagent volume of 80mL. Jars are constructed of break-resistent acetal polymer, and are resistant to most staining agents including alcohol and xylene, but not phenol.
  • Jars can be loosely joined to each other to keep the same order when moved around
  • Wide stable base for stability while inside is recessed to reduce reagent volume
  • Available in five colours to aid identification of contents or applications
The slide staining rack holds up to 12 microscope slides of varying dimensions. Slides fit into individual slots for free passage and rapid drainage of staining fluids. Due to their vertical insertion within the rack, the slides writing area will not be stained by the fluid, allowing the slides to be removed without the use of forceps.
The staining rack is also constructed of acetal polymer and is specially formulated for rapid drying in a microwave oven, or at temperatures up to 120°C (248°F). The lid completely covers the EasyDip™ slide staining jar to minimize spills or evaporation. A handle is permanently attached to the rack for easy insertion and removal.
Ordering information: The staining jars and staining rack are sold separately. The EasyDip™ Kit with Rack includes one anodized aluminum holder along with six assorted color jars (two white ones) and one M905-12DGY Slide Staining Rack. The EasyDip™ Aluminum Holder is designed to hold six EasyDip™ slide staining jars. The assorted kit includes one jar of each color.
Product Family Options
Información del producto
  • Dimensions
    Color
    Availability
    Price
  • 87000-120
    EasyDip™ Slide Staining Jars
    EasyDip™ Slide Staining Jars
    Slide Staining Jars: 64L x 76W x 92H mm (21/2 x 3 x 35/8"), Slide Staining Rack: 60L x 64W x 97H mm (23/8 x 21/2 x 313/16")
    Blue
    Log in Para disponibilidad
    Log in Para disponibilidad
    Case (6UN)
    Case (6UN) - $2,036.64
    $2,036.64
    Case (6UN)
       $2,036.64
    Supplier Number: M900-12B
  • 87000-132
    EasyDip™ slide staining rack
    EasyDip™ slide staining rack
    60L x 64W x 97H mm (23/8 x 21/2 x 313/16")
    Dark grey
    Log in Para disponibilidad
    Log in Para disponibilidad
    Case (6UN)
    Case (6UN) - $2,516.01
    $2,516.01
    Case (6UN)
       $2,516.01
    Supplier Number: M905-12DGY
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 