Harvard Trip Balances, Ohaus
12351-024
- Descripción:Double Beam with 225 g Tare, Stainless Steel Plates
- Referencia:12351-024
- Tiempo de estabilización:1.5 s
- Legibilidad:0.1 g
- Capacidad de pesaje:2000 g (4.4 lbs.)
- Nº de proveedor:80000005
Versatile, rugged, easy-to-use balances are ideal for comparative weighing
- 10 x 0.1 g (Upper Beam), 200 x 10 g (Lower Beam)
The footed polypropylene scoop with counterweight may be ordered separately.
Balances feature angled beam with center reading poise, magnetic damping, zero adjust compensator, and precision ground knives. Stainless steel 15.2cm (6") weighing platform. Balance 12351-024 includes a 225g tare beam.
: Supplied with manufacturer’s five-year warranty.