ContactUsLinkComponent
Valor® 2000 Compact Industrial Bench Scale
Valor® 2000 Compact Industrial Bench Scale
  10123-890
cUL Listed
CSA Certified
UL Listed
undefined
Valor® 2000 Compact Industrial Bench Scale
  10123-890
 :  30035441
cUL Listed
CSA Certified
UL Listed
  • Modelo:
    V22PWE6T
  • Tamaño de la plataforma/la charola:
    190×242 mm (7.5×9.5")
  • Dimensiones:
    256×280×121 mm
  • Rango de temperatura de funcionamiento:
    –10...+40 °C (14...104 °F)
  • Referencia:
    10123-890
  • Linealidad:
    ±1 g
  • Tiempo de estabilización:
    ≤0.5 s
  • Legibilidad:
    0.001 kg (0.002 lb.)
  • Capacidad de pesaje:
    6000 g (15 lbs.)
  • Nº de proveedor:
    30035441
High-performance food scale suitable for harsh, wet workplace conditions. The sturdy polycarbonate keypad offers protection from knives and other sharp instruments used in food preparation, processing and portioning applications.
  • IPX8 flow-thru design ensures that the NSF-certified Valor® 2000 will drain moisture in the scale when used in wet conditions
  • Stabilization time is less than a half-second
  • Large front and rear LED displays allow easy viewing so two operators can share one scale
The OHAUS Valor 2000 water resistant food scale raises the bar and sets a new industry standard with the winning combination of speed and durability. Available in both stainless steel and plastic housing designs, Valor® 2000 offers large LED front and rear displays, food safety and puncture resistant keypads, making it the premier value scale for use in dry or washdown environments.
Power: 12 V DC 0.84 A, AC Universal adapter (100 - 220 V AC / 50 - 60 Hz) and built-in internal rechargeable lead acid battery included.
 : In compliance with DIN ISO 4796-1 standards.