Orders LinkContactUsLinkComponent
Soder-Wick® Desoldering Braids, ITW Chemtronics®
Soder-Wick® No Clean SD on Static-Dissipative Bobbins, 1.5 m (5'), 2.8 mm (0.11")
  15145-658
 :  80-4-5
Soder-Wick® Desoldering Braids, ITW Chemtronics®
  15145-658
Soder-Wick® Desoldering Braids, ITW Chemtronics®
Soder-Wick® Desoldering Braids, ITW Chemtronics®
Soder-Wick® No Clean SD on Static-Dissipative Bobbins, 1.5 m (5'), 2.8 mm (0.11")
$4,801.79
Paquete de 25
Desoldering braids significantly reduce rework/repair time and minimize the risk of heat damage
Soder-Wick® Rosin is a Type R flux braid that is fast, safe, noncorrosive, and leaves no ionic contamination.
Soder-Wick® No Clean SD braid is noncorrosive, halide-free, and organic.
Meet Bellcore TR-NWT-000078 and IPC SF-818, and ANSI/IPC J STD-004, Type ROL0.
The weave design allows maximum solder capacity and optimizes heat transfer. Braids leave little or no flux residue, shortening or completely eliminating cleaning time.
 : Soder-Wick® Rosin flux is available in either ESD-safe static-dissipative bobbins sealed in Performance Pak™ barrier packaging (25 bobbins per pack) or in individual standard spool packaging. Soder-Wick® No Clean SD flux is available on static-dissipative bobbins that are sealed in Performance Pak™ barrier packaging (25 bobbins per pack).
  • Width:
    2.8 mm (0.11")
  • Description:
    Soder-Wick® No Clean SD on Static-Dissipative Bobbins
  • Length:
    1.5 m (5')