VWR® Plate Roller
VWR® Plate Roller
Supplier: VWR International
Jump to:
Product Details & Documents
This 10.2 cm (4") soft rubber roller is designed for assuring tight and consistent application of adhesive films and foils to microplates.
- Tight and consistent application
- Secure seal around all microplate wells
Documents
Especificaciones
- Description:VWR® Plate Roller