Orders LinkContactUsLinkComponent
Slide Drying Bench, Electrothermal
Slide Drying Bench, Electrothermal
Cole-Parmer® SW-200-115 Microscope slide warmer, 115 V, 50 (1× 3"), 15.5W×7.12L×3.5"H (39.37W×18.09L×8.89H cm), 100 °C
Catalog # 15156-120
Supplier:   ANTYLIA SCIENTIFIC
Número de proveedor #:  48573-01
Slide Drying Bench, Electrothermal
Catalog # 15156-120
Supplier:  ANTYLIA SCIENTIFIC
Slide Drying Bench, Electrothermal
Slide Drying Bench, Electrothermal
Cole-Parmer® SW-200-115 Microscope slide warmer, 115 V, 50 (1× 3"), 15.5W×7.12L×3.5"H (39.37W×18.09L×8.89H cm), 100 °C
Price
$31,756.18
Log in Para disponibilidad
Each (1UN)
Product Details & Documents
For drying slides at the mounting stage in histology applications.
  • Temperature range from ambient to 100 °C
  • Drying bars included for easier slide drying
  • Arrange slides in many different orientations
  • Dark surface makes it easy to see samples
For 230V model, contact your VWR sales representative.
Equipped with built-in energy regulator (for AC only).
Delivery information: Supplied with four support rods and power cord with US plug.
Documents
Especificaciones
  • Temperature Range:
    100 °C
  • Capacity:
    50 (1× 3")
  • Description:
    Cole-Parmer® SW-200-115 Microscope slide warmer
  • Dimensions:
    15.5W×7.12L×3.5"H (39.37W×18.09L×8.89H cm)
  • Electrical:
    115 V