ContactUsLinkComponent
1860 Disposable N95 Health Care Particulate Respirator and Surgical Masks 3M™
1860 Disposable N95 Health Care Particulate Respirator and Surgical Masks 3M™
Catálogo # 43100-064
Proveedor:  Healthcare Products
undefined
1860 Disposable N95 Health Care Particulate Respirator and Surgical Masks 3M™
Catálogo # 43100-064
Proveedor:  Healthcare Products
Número de proveedor:  MMM 1860
Especificaciones
  • Tamaño:
    Regular
  • Referencia:
    43100-064
  • Nº de proveedor:
    1860
Especificaciones
Acerca de este artículo
This respirator has a soft inner shell for extended comfort on the face.
  • Fluid-resistant to reduce the potential exposure of the wearer to blood and other body fluids
  • Provides minimum filtration efficiency of 95% against certain solid and liquid particles that do not contain oil
Meets CDC guidelines for TB exposure control and helps reduce exposure to airborne particles.
Precaución: All 3M™ products must be used in accordance with OSHA regulations and the user instructions, warnings, and limitations accompanying each product.