Tubing and Dispensing Accessories, WHEATON®, DWK Life Sciences
Supplier: DWK LIFE SCIENCES S. DE R.L. DE MX
Certificates
About this item
A wide variety of tubing for peristaltic pumps and liquid handling needs.
Peroxide Cured Silicone, USP Class VI Silicone, Thermo Plastic Elastomer(TPE) or Black Synthetic Rubber (FKM).
Related Information
Frequently Bought Together