Barnstead/Lab-Line Universal Platforms for Shakers, Thermo Scientific
Catalog # 97040-486
Supplier: Thermo Fisher Scientific
Specifications
- Dimensiones:45.7L x 45.7W cm (18 x 18")
- Referencia:97040-486
- Para ser utilizado con:MaxQ™ 2000, MaxQ™ 6000 Shaker
- Nº de proveedor:30128
Specifications
About this item
Platforms may be customized for specific shaking requirements
Platforms are drilled and tapped with the maximum number of holes for use with a variety of different types of vessels.
Ordering information: Platforms only. For clamps, see the Accessories for MaxQ™ 8000 Stackable Orbital Shakers, Thermo Scientific series. For tube racks for use with universal platforms, see the Barnstead/Lab-Line Unwire™ Test Tube/Microcentrifuge Tube Racks for Universal Platforms, Thermo Scientific series.