Mantle Extension Support, Series M, Glas-Col®
33781-043
: GLAS-COL
Small Business Enterprise
- Capacidad:250 - 500 ml
- Referencia:33781-043
- Diámetro interno:16.2 cm (6⅜")
- Nº de proveedor:100EM534
Steel ring with 15.2 cm (6") shank. Gray finish support mantles to 5 L.
Attaches to ring stand or distillation rack
Adjusts to convenient height