VWR® Nutating Mixer
115 V, 60 Hz, 24 rpm
Catalog # 82007-202
Supplier: VWR International
Jump to:
Product Details & Documents
The three-dimensional shaking motion of this mixer is gentle yet efficient, making it ideal for mixing samples in blood and centrifuge tubes.
- Three-dimensional motion
- Provides thorough yet gentle mixing
- Ideal for blood tubes and centrifuge tubes
Mixer features a steel platform. Permanently lubricated bearings on moving parts provide quiet, maintenance-free operation. Speed and tilt angle are fixed to provide the optimal level of agitation for mixing samples in small containers without foaming. Mixer can be used in a cold room. A small footprint allows the mixer to be placed on a crowded bench.
Ordering information: Supplied with a dimpled rubber mat for firmly holding blood collection tubes, microtubes, or 15 and 50 ml centrifuge tubes.
Documents
Especificaciones
- Electrical:115 V, 60 Hz
- Speed:24 rpm
- Shipping weight:1 kg (2.2 lbs.)
- Cat. No.:82007-202
- Platform dimensions:24.8W×17.2D cm (9³/₄×6³/₄")
- Tilt angle:20°, fixed
- Platform size:19×12.7 cm (7¹/₂× 5")
- Max. load:0.8 kg (1.8 lbs.)
- Overall dimensions:15.2 cm (6") (Height)
- Operating temperature:4...40 °C (39.2...104 °F)